Proto® Internal Pipe Wrenches, Essendant
Catalog # 76189-312
Supplier: Safety & Industrial Supplies
Specifications
- Material:Steel
- Cat. No.:76189-312
- Overall length:3"
- Finish:Black oxide
- Opening size:⅜"
Specifications
About this item
Ideal for removing broken threaded pipe ends pipe plugs and fittings.
- Installation and removal without thread damage
- Eccentric cam action engages wrench against the walls of the pipe
- Hex body allows for use of adjustable wrench for added torque
- Made from hardened steel and plated to help prevent rust