ContactUsLinkComponent
Proto® Tether-Ready Soft Jaw Cannon Plug Pliers, ORS Nasco
Proto® Tether-Ready Soft Jaw Cannon Plug Pliers, ORS Nasco
Catalog # 76190-620
Proto® Tether-Ready Soft Jaw Cannon Plug Pliers, ORS Nasco
Catalog # 76190-620
Supplier Number:  577-253G-TT
Specifications
  • Length:
    9½"
  • Cat. No.:
    76190-620
  • Jaw type:
    Soft, curved
Specifications
About this item
Replaceable urethane padded jaws protect work piece from marking or scratching.
  • Key-ring tether attachment point in handle
  • Cutting capacity of 3/4" to 21/2"
  • Four-position pivot design provides greater concentricity
  • Plastisol grips for added comfort