Order Entry
United States
ContactUsLinkComponent
Gas Inlet Adapter, with Hose Connection, Chemglass
Gas Inlet Adapter, with Hose Connection, Chemglass
Catalog # 80066-118
Supplier:  Chemglass
Small Business Enterprise
undefined
Gas Inlet Adapter, with Hose Connection, Chemglass
Catalog # 80066-118
Supplier:  Chemglass
Supplier Number:  CG-1060-01
Small Business Enterprise

Specifications

  • Description:
    Gas Inlet Adapter, with Hose Connection
  • Cat. No.:
    80066-118
  • [ST] joint size:
    24/40
  • Supplier no.:
    CG-1060-01

Specifications

About this item

Gas inlet adapter with a tube extending 200 mm below the inner joint.

  • 24/40 joint size
  • With serrated hose connection having an O.D. of 10 mm at the largest serration

Permits gas to be introduced at the bottom of larger reaction vessels or flasks.