Connecting Adapter, with 105° Side Outer Joint, Chemglass
Catalog # 80065-828
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Connecting Adapter, with 105° Side Outer Joint
- Cat. No.:80065-828
- [ST] joint size:24/40
- Supplier no.:CG-1023-01
Specifications
About this item
Adapter features standard taper joints at top and side, with inner joint at bottom.
- 24/40 joint size
- Side outer joint at 105°
- Manufactured by Chemglass
Sidearm is positioned at a 105° angle from the lower tube.