Inert Gas Inner Joint Adapters, Chemglass
Adapters have two hose connections 180° from one another to permit a flow of inert gas over the apparatus.
Hose connection has an O.D. of 10 mm at the largest serration.
Frequently Bought Together
Adapters have two hose connections 180° from one another to permit a flow of inert gas over the apparatus.
Hose connection has an O.D. of 10 mm at the largest serration.