ContactUsLinkComponent
arium® UV Lamp (185/254 nm), Sartorius Corp.
  75803-118
 :  Sartorius
arium® UV Lamp (185/254 nm), Sartorius Corp.
  75803-118
 :  Sartorius
 :  H2O-CEL1
  • Description:
    arium® mini UV Lamp (185 | 254 nm) for arium® mini, arium® mini essential and arium® mini plus
  • Cat. No.:
    75803-118
  • Supplier no.:
    H2O-CEL1
Ultrapure water free of TOC.
  • Horizontal installation; optimized temperature gradient
  • Effective breakdown of organic compounds
  • Prevents the growth of microorganisms
  • Easy replacement
  • TOC for purified water* <5 ppb
Horizontally aligned, the UV lamp delivers especially reliable results. Unlike vertical units, the temperature gradient is less pronounced and therefore does not affect the activity of UV radiation. With two different wavelengths, the UV lamp reliably removes total organic compounds (TOCs), efficiently preventing microbial growth. At 185 nm, organic compounds are oxidized and at 254 nm, microorganisms are killed off.