Aven In-X Interchangeable Magnifying Lamp with 5 Diopter Lens, Aven
Catalog # 76289-588
Supplier: Aven Tools
Specifications
- Magnification:2.25×
- Description:Interchangeable Magnifying Lamp
- Cat. No.:76289-588
Specifications
About this item
Cutting edge lamp designed to expand magnification range by accommodating multiple lenses.
- Easy lens exchange: no tools required
- Eliminate multiple magnifying lamps
- Rotatable lamp head
Capable of a magnification of 2.25 to 4.75× with the optional lenses.