About this item
These polystyrene reagent reservoirs offer a convenient solution for the temporary storage of non-solvent based liquids during any pipetting application.
- Integrated volume graduations result in more accurate measurements and less reagent waste
- Convenient pour back spout is molded into each reagent reservoir, allowing precise control of liquid flow
- Multichannel reagent reservoirs feature an extra deep trough that accumulates liquid, reducing dead volumes and ensuring maximum reagent recovery
Each reusable reservoir base securely holds disposable clear reservoir inserts during use. Reservoir inserts neatly nest inside one another, significantly reducing storage requirements. Plastic waste is also reduced by only replacing the insert and not an entire reservoir.
Reusable Reservoir Bases: A sturdy, reusable base is included with every case of bulk sealed reservoirs. The multichannel reservoir base is designed for standalone use, or with the VIAFLO ASSIST and ASSIST PLUS.
The 10, 25 and 100 ml multichannel reservoir bases can accommodate a second reservoir insert as a lid, preventing evaporation or contamination.