Jess or Wes Separation Module, Protein Simple
Catalog # MSPP-SMW004
Supplier: PROTEIN SIMPLE MS
CAS Number:
Specifications
- Description:Jess or wes separation module
- Cat. No.:MSPP-SMW004
Specifications
About this item
Reagents for 200 data points to use with a detection module.
Includes two parts: Separation box with 8 Capillary Cartridges (25 capillaries per cartridge), 8 Jess/Wes 12-230 kDa Pre-filled Plates (pre-filled with Separation Matrix 2, Stacking Matrix 2, Split Running Buffer 2 and Matrix Removal Buffer), Sample Buffer, Wash Buffer (Store at RT), plus Standard Pack 1 clamshell with 8 ea. DTT, FL Standard master mix with 29 kDa System Control protein. (Store at 4¡C).