About this item
These micronic TPE multi pierceable capcluster are used to seal 96 individual tubes.
- Sterile, gamma radiated (15 kGy) or EtO treated
- Pre-capped on micronic tubes
- Split septum to pierce with a pipette tip
- Resists high chemical solvents
- Temperature range: –80 to +100 °C
- Easy visual differentiation with different colors
The Capcluster-96 is a sheet of 96 push caps that can be used to seal an entire rack of 96 tubes at once. After the capcluster is used to seal a rack of push cap tubes, the foil backing can be removed to leave each tube individually capped. These capclusters can be pierced repeatedly without losing seal integrity. They can be used to seal sample storage tubes in temperatures as low as -80°C.
All caps are produced in a class 7 clean room and are RNase/DNase and Pyrogen free.