Order Entry
United States
ContactUsLinkComponent
Stainless Steel ATD Prepacked Sample Tubes, PerkinElmer
Stainless Steel ATD Prepacked Sample Tubes, PerkinElmer
Catalog # 10136-870
Supplier:  PerkinElmer
undefined
Stainless Steel ATD Prepacked Sample Tubes, PerkinElmer
Catalog # 10136-870
Supplier:  PerkinElmer
Supplier Number:  N9307002

Specifications

  • Description:
    ATD prepacked sample tubes
  • Material:
    Stainless steel
  • Cat. No.:
    10136-870
  • For use with:
    Carbopack™ B 60/80 sorbent
  • Supplier no.:
    N9307002

Specifications

About this item

These prepacked sample tubes are designed for Carbopack™ B 60/80.

  • Analyte volatility range is n-C5 to n-C12

The Carbopack™ B is conditioned at 350 °C. These tubes are deigned with maximum temperature of 400 °C with a desorption temperature of 330 °C.