Order Entry
United States
ContactUsLinkComponent
SSC Buffer, 20X Concentrate, Seracare
SSC Buffer, 20X Concentrate, Seracare
Catalog # 95059-206
Supplier:  KPL
CAS Number:  
SSC Buffer, 20X Concentrate, Seracare
Catalog # 95059-206
Supplier:  KPL
Supplier Number:  5960-0021
CAS Number:  

Specifications

  • Description:
    20X SSC Buffer
  • Size:
    1 l
  • Cat. No.:
    95059-206
  • pH:
    7.0
  • Supplier no.:
    50-86-05

Specifications

About this item

KPL 20X SSC is a common buffer used in a variety of molecular biology applications.

  • Clear, colorless solution
  • Made up of molecular biology-grade 3.0 M NaCl and 0.3 M sodium citrate

The solution is filtered through a 0.2 filter into a DNase/RNase-free bottles. KPL 20X SSC can be used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts.