BD Difco™ Salmonella Antiserum O Vi, BD Diagnstics
Catalog # 90001-986
Supplier: BD
CAS Number:
Specifications
- Description:Difco™ Salmonella antiserum O Vi
- Size:3 ml
- Cat. No.:90001-986
- Supplier no.:228271
Specifications
About this item
BD offers antigens and antisera that can facilitate diagnosis of several diseases and meet your microbiology needs.
- Used in slide agglutination tests for the identification of Salmonella Vi
BD Difco™ Salmonella Vi Antisera are lyophilized, polyclonal rabbit antisera containing approximately 0.04% Thimerosal as a preservative. When properly rehydrated and used as recommended, each vial of BD Difco™ Salmonella Vi Antisera contains sufficient reagent for 60 tests. BD Difco™ Salmonella Vi Antisera are prepared with representative strains of these serogroups and are not absorbed.