ContactUsLinkComponent
ICP Calibration Standard, Surface Water, Inorganic Ventures
ICP Calibration Standard, Surface Water, Inorganic Ventures
Catalog # 103255-014
Supplier:  Inorganic Ventures
CAS Number:  
ICP Calibration Standard, Surface Water, Inorganic Ventures
Catalog # 103255-014
Supplier:  Inorganic Ventures
Supplier Number:  IV-STOCK-10-125
CAS Number:  
Specifications
  • Description:
    ICP Calibration Standard-Surface Water
  • Size:
    125 ml
  • Cat. No.:
    103255-014
  • Supplier no.:
    IVSTOCK-10-125
Specifications
About this item
ICP calibration standard surface water is a 125 ml certified reference material set in a nitric acid matrix for stability.
  • 23 analytes
Certifications: ISO 17025 and ISO 17034 certified.