ContactUsLinkComponent
Microscope Slide Mailer, Ted Pella
Microscope Slide Mailer, Ted Pella
Catalog # MSPP-22519
Supplier:  TED PELLA INC MS
Microscope Slide Mailer, Ted Pella
Catalog # MSPP-22519
Supplier:  TED PELLA INC MS
Supplier Number:  22519
Specifications
  • Color:
    Black
  • Description:
    Microscope Slide Mailer
  • L×W:
    3×1"
  • Cat. No.:
    MSPP-22519
  • Supplier no.:
    22519
Specifications
About this item
This convenient disposable staining container is also useful for shipping, storing and handling of slides.
  • Made of polypropylene
  • Container holds five slides placed without touching each other
  • Positive lock and flip-top