Order Entry
United States
ContactUsLinkComponent
Mighty Vue Pro Magnifying Lamp, Aven Tools
Mighty Vue Pro Magnifying Lamp, Aven Tools
Catalog # 76251-612
Supplier:  Aven Tools
undefined
Mighty Vue Pro Magnifying Lamp, Aven Tools
Catalog # 76251-612
Supplier:  Aven Tools
Supplier Number:  26505-ESL-XL5

Specifications

  • Magnification:
    2.25×
  • Color:
    Black
  • Description:
    MightyVue Pro LED Magnifying Lamp
  • Cat. No.:
    76251-612
  • W×D:
    7.5×6.2"
  • Diopters:
    5

Specifications

About this item

Aven’s Mighty Vue Pro LED series magnifying lamps are in a class of their own. Featuring adjustable color temperature (3500 to 6500 K), brightness control (max 1100 lm), in addition to an attractive design, the Mighty Vue Pro series lamps are the most advanced available.

  • Touch-sensitive brightness and color temperature controls
  • 36" spring-balanced arm ensures a large working range
  • 360° swivel motion allows the head to be placed in any position
  • Covered spring arm prevents pinching hazards
  • ESD safe coating
  • 60 LED lights