The RapidPURE™ Plasmid mini kit is designed to isolate up to 20 μg of plasmid DNA from 1.5 ml of an overnight bacterial culture for use in routine molecular biology applications such as fluorescent and radioactive sequencing and cloning.
- Save time with turbo filter design
- Process with either vacuum or centrifugation
- Plasmid DNA ready for transfection and other downstream applications
- No phenol/chloroform extraction or precipitation required
- Use with CIRCLEGROW® Broth for even higher yields
The RapidPURE™ Plasmid Mini Kit is designed to isolate up to 20 μg of plasmid DNA from 1.5 ml of an overnight bacterial culture. All steps can be carried out in either a microcentrifuge or vacuum manifold.
The Turbo Filter is combined with a catch tube into a Turbo Cartridge featuring one hole in the center of the cap for the in-situ addition of wash solution, and a second hole permitting in-situ removal of waste solution using vacuum aspiration.
Application includes: Nucleic Acid Isolation and Purification; Sample Lysis and Preparation.
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Storage Conditions:Store at Room temperature (15 - 30 °C).
- Description:Kit and Assay