SD Medium-Leu-Trp (Pouches), MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Double dropout formulation of synthetic defined (SD) yeast media which includes all necessary components for selective growth of S. cerevisiae. SD and SD with agar (SDA) in a variety of formats for flexibility and convenience.
Application includes: Yeast Cell Culture, Gene Expression.
Specifications
- Storage Temperature:RT
Specifications
Frequently Bought Together