SOB Medium, MP Biomedicals
76202-286
:
- Description:Powdered Bacterial Media
- Size:454 g (1 lbs.)
- Storage Temperature:RT
- Cat. No.:76202-286
SOB Medium Rich Medium for the preparation of competent host cells prior to transformation. Certified for plasmid and other bacterial cultures in liquid medium. Standard media for preparing cells for chemical transformation.
- Standard media for preparing cells for chemical transformation
- 31 g (31 capsules) per liter
- Contents/l: 20 g tryptone, 5 g yeast extract, 0.5 g NaCl, 5 g MgSO₄·7H₂O
Biological buffers have been added for proper pH adjustment. Gelatin capsules are produced in a facility that meets the FDA’s recent guidance as being derived from BSE-free herds that are born, raised, and slaughtered in the United States.
Application includes: Bacteria Cell Culture, Transformation.
: Molecular Biology Certified bacterial growth medium.