Order Entry
United States
ContactUsLinkComponent
NZCYM Broth Capsules, MP Biomedicals
NZCYM Broth Capsules, MP Biomedicals
Catalog # 76202-468
Supplier:  MP Biomedicals
CAS Number:  
NZCYM Broth Capsules, MP Biomedicals
Catalog # 76202-468
Supplier:  MP Biomedicals
Supplier Number:  113004031
CAS Number:  

Specifications

  • Description:
    Bacterial Media Capsules
  • Size:
    1 kg (2.2 lbs.)
  • Storage Temperature:
    RT
  • Cat. No.:
    76202-468

Specifications

About this item

Rich medium for the growth of lambda phage in E. coli in liquid medium.

  • Proprietary media for enhanced cell yield
  • Convenient packaging formats for a variety of applications and budgets
  • Proven and reliable reputation
  • High-quality raw materials and additives
  • Encapsulated media
  • No weighing, dust and cleanup
  • Drop capsules into water and autoclave

23 g (23 Capsules)/l. Contents/l: 10 g NZ-amine, 1 g casamino acids, 5 g yeast extract, 2 g MgSO₄·H₂O, 5 g NaCl, pH 7.

Application includes: Bacteria Cell Culture, Microbiological Analysis, Gene Expression.

Certifications: Molecular Biology Certified™.