Order Entry
United States
ContactUsLinkComponent
LB Medium, Lennox Capsules, MP Biomedicals
LB Medium, Lennox Capsules, MP Biomedicals
Catalog # 76019-956
Supplier:  MP Biomedicals
CAS Number:  
undefined
LB Medium, Lennox Capsules, MP Biomedicals
Catalog # 76019-956
Supplier:  MP Biomedicals
Supplier Number:  113002131
CAS Number:  

Specifications

  • Description:
    Bacterial Media Capsules, Acceptable pH range is 6.9 ±0.3 at 25 °C
  • Size:
    1 kg (2.2 lbs.)
  • Storage Temperature:
    Store sterile LB Agar, Lennox at ambient temperature (15 - 30 °C). Powder medium is hygroscopic, therefore minimize exposure to air to prolong shelf life of media.
  • Cat. No.:
    76019-956

Specifications

About this item

Luria-Bertani, Rich Medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.

  • Proprietary media for enhanced cell yield
  • Convenient packaging formats for a variety of applications and budgets
  • Proven and reliable reputation
  • High-quality raw materials and additives
  • Encapsulated media
  • No weighing, dust and cleanup
  • Drop capsules into water and autoclave

LB medium lennox powder is a nutritionally rich medium originally developed for growth and maintenance of E. coli strains. The growth characteristics are the same as L-Broth Agar powder. L-Broth supports a luxurious growth of E. coli cells as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesize. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.

Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis, protein expression.

Certifications: Molecular Biology Certified™. Gelatin capsules are produced in a facility that meets the FDA’s recent guidance as being derived from BSE-free herds that are born, raised, and slaughtered in the United States.