LB Medium Powder, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.
- Proprietary media for enhanced cell yield
- Convenient packaging formats for a variety of applications and budgets
- Proven and reliable reputation
- High-quality raw materials and additives
LB Medium is a nutritionally rich medium originally developed for growth and maintenance of E. coli strains. LB Medium supports a luxurious growth of E. coli cells as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesize. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.
Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis, protein expression.
Certifications: Molecular Biology Certified™.
Specifications
- Storage Temperature:Store sterile LB Medium at ambient temperature (15 - 30 °C). Powder medium is hygroscopic, therefore minimize exposure to air to prolong shelf life of media.
Specifications
Frequently Bought Together