Order Entry
United States
ContactUsLinkComponent
FastPROTEIN Blue, MP Biomedicals
FastPROTEIN Blue, MP Biomedicals
Catalog # 76020-014
Supplier:  MP Biomedicals
CAS Number:  
undefined
FastPROTEIN Blue, MP Biomedicals
Catalog # 76020-014
Supplier:  MP Biomedicals
Supplier Number:  116550500
CAS Number:  

Specifications

  • Description:
    FastProtein™ Blue Matrix
  • Size:
    100×2 ml tubes
  • Cat. No.:
    76020-014

Specifications

About this item

The FastPROTEIN blue kit is designed for rapid isolation and purification of protein from bacterial cultures.

  • Easy lysis of bacterial cells to release recombinant proteins
  • DNase and RNase-free lysing matrix tubes

The FastPROTEIN™ Blue Matrix, also known as Lysing Matrix B, is used with any FastPrep® Instrument to reproducibly lyse gram positive and gram negative bacterial cells within 40 seconds. Rapid release of recombinant protein saves valuable time during optimization of expression.

Information from up to 24 different induction variables can be quickly obtained after one 40 second FastPrep® run followed by electrophoresis. Once your protein of interest is expressed and identified, the active proteins released by the unique motion of the FastPrep® Instruments can be recovered from the lysate by a wide range of protein-specific techniques.

Application includes: Sample preparation; sample lysis; nucleic acid isolation and purification; molecular biology; gene expression; microbiological analysis; gene function; gene regulation.

Ordering information: FastPrep® Instrument or alternative high speed disruptor, empty lysing matrix tubes and caps, microcentrifuge, pipettmen, purification reagents.
Caution: Store at room temperature (15 - 30 °C)