ContactUsLinkComponent
Infrared Heating Lamp, Cole Parmer
Infrared Heating Lamp, Cole Parmer
Catalog # MSPP-EW0305700
Supplier:  Cole Parmer
undefined
Infrared Heating Lamp, Cole Parmer
Catalog # MSPP-EW0305700
Supplier:  Cole Parmer
Supplier Number:  03057-00
Specifications
  • Description:
    Infrared Heating Lamp
  • Cat. No.:
    MSPP-EW0305700
  • Electrical:
    115 V, 50/60 Hz
  • Supplier no.:
    EW-03057-00
Specifications
About this item
Lamp can be mount to any table or benchtop, or use our floor stands to support the heating lamp.
  • Features durable all-metal construction
  • Dual reflector prevents excessive heat buildup
Position of the lamp can be adjusted using 42" swiveling extension arm. Lamp comes complete with screw-type mounting bracket, 10-ft grounded cord with plug and 250 watt, hard glass infrared heat bulb. Easily remove wire bulb guard to replace bulb.
Ordering information: Replacement bulb should be ordered separately.