Cartridge Blade 10 S1, JBC
Catalog # 76241-410
Supplier: JBC TOOLS USA INC. TE
Specifications
- Description:Cartridge Blade
- Cat. No.:76241-410
Specifications
About this item
This cartridge has a 10 mm wide blade tip.
C245 cartridge range is the widest, with many shapes, sizes and special models. They work with T245 handpieces.
They are appropriate for general soldering purposes and they are perfect for both SMD components or for applications with high power requirements such as ground plane soldering.