Dual-Wire Fabric Wristband, SCS, Desco Industries
Supplier: Desco
Certificates
About this item
The Dual-Wire Fabric Wristband is available in one size that adjusts to almost any size wrist for reliable protection.
- Adjustable, elastic nylon material with conductive woven thread
- Fits most wrist sizes to provide reliable protection
- Dual-Wire conductor provides redundancy
- Even if one dual-wire wrist strap conductor is severed, operator has reliable path-to-ground with other wire
The wristband provides full conductivity, comfort, and reliability, while ensuring continuous drain of static charge when used with SCS 2300 Dual-Wire Coil Cords (sold separately). Made to be used in conjunction with all SCS Workstation Monitors.
Certifications: Meets requirements of ANSI/ESD S20.20 and ANSI/ESD S1.1.
Frequently Bought Together