ContactUsLinkComponent
Disposable N95 Particulate Respirators, 4800, Moldex®
Disposable N95 Particulate Respirators, 4800, Moldex®
Catalog # 10040-388
Supplier:  Moldex/Metric
   Safer Human and Environmental Health
undefined
Disposable N95 Particulate Respirators, 4800, Moldex®
Catalog # 10040-388
Supplier:  Moldex/Metric
Supplier Number:  4800
   Safer Human and Environmental Health
Specifications
  • Description:
    N95 disposable respirators
  • Size:
    Medium/large
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Cat. No.:
    10040-388
  • Supplier no.:
    4800
Specifications
About this item
The N95 disposable respirator has added carbon layer helps relieve workers of nuisance levels (Below the OSHA Permissible Exposure Limit) of ozone and organic vapor odors.
  • Suitable for most welding fumes
  • Fits easily under welding hoods
  • Less resistance, better breathability
  • 100% PVC-free
SmartStrap® allows the AirWave® to hang from the neck when not in use. Softspun® lining for increased comfort and durability. Its adjustable soft cloth strap feels cool and comfortable. Full foam face seal for added comfort and no pressure points and metal-free.
Dura-Mesh® shell protects the filter media so it stays cleaner looking longer and resists collapsing in heat and humidity.
Certifications: Meets heat and flame resistance in accordance with ANSI/ISEA 110-2003 Section 7.11.1. Certified under 42CFR84.