VWR® Sport Safety Glasses
Catalog # 76210-730
Supplier: VWR International
Specifications
- Frame color:Smoke
- Cat. No.:76210-730
- Lens coating:Anti-fog, scratch-resistant
- Lens tint:Clear
Specifications
About this item
Wrap-around protective glasses combine fashion, fit, and function.
- Anatomically-shaped nose adapts to all faces
- Co-injected comfort pad will not peel off in hot climates
- High-visibility rubber color
- Polycarbonate lens
- Provides 99.9% UV protection
Inner temple coated with elastomer for excellent grip and soft cushioning with impact-resistance.
Certifications: Meets ANSI Z87.1 standards.