Prostaglandin Screening Library I, Cayman Chemical
Catalog # 76271-072
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Description:Prostaglandins and Isoprostanes in a 96-well Matrix Tube rack format
- Cat. No.:76271-072
- No. of wells:96 wells
Specifications
About this item
For screening of PGF analogs
- Contains >70 compounds
- Includes the “F-series” prostaglandins and isoprostanes
The Prostaglandin Screening Library I contains more than 70 “F-series” of prostaglandins and isoprostanes in a 96-well Matrix tube rack format as 2 mM stocks in DMSO. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.