Tech-Med® Sprague Rappaport Type Stethoscopes, Dukal
Supplier: Dukal
Certificates
About this item
Stethoscopes provides versatile acoustical response for infants, children, and adults.
- PVC dual-channel tubing, chrome plated brass chest piece, and binaural
- Small, medium, and large bells screw into chest piece to detect low to medium frequency sounds
- Complete with accessory pouch, ID tag, and instruction manual
- Lifetime warranty
Frequently Bought Together