ContactUsLinkComponent
Tech-Med® Dual Scale Digital Thermometer, DUKAL Corporation®
Tech-Med® Dual Scale Digital Thermometer, DUKAL Corporation®
Catalog # 76235-462
Supplier:  Dukal
undefined
Tech-Med® Dual Scale Digital Thermometer, DUKAL Corporation®
Catalog # 76235-462
Supplier:  Dukal
Supplier Number:  3210D-360
Specifications
  • Description:
    Dual Scale Digital Thermometer
  • Cat. No.:
    76235-462
Specifications
About this item
Built-in memory enables display of previously measured values.
  • Quick 10 second read with easy to read, large LCD display and low battery indication
  • Measurement range: 32...43.9 °C, 90...109.9 °F
  • Alarm/beeper sounds when reading is complete
  • 200 hours of continuous battery powered operation
  • Auto shut off after 9 minutes of non-use