




Sulfuric acid : Phosphoric acid blend (1:1)
Catalog # RCR8347000-4A
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:4 L
- Cat. No.:RCR8347000-4A
Specifications
Specification Test Results
- Appearance:Colorless liquid
Specification Test Results
About this item
:
UN:3264
ADR:8,II
:GHS05
:GHS08
:GHS06
:GHS09
:GHS07
Storage Temperature:Ambient