UNEX Buffer, Microbiologics
Catalog # 76288-260
Supplier: MICROBIOLOGICS, INC.
CAS Number:
Specifications
- Description:UNEX Buffer
- Cat. No.:76288-260
Specifications
About this item
Ready-to-use UNEX Buffer is an isothiocyanate lysis buffer specially designed for preserving samples and universal nucleic acid extraction (UNEX) of DNA and RNA from viruses, bacteria, and parasites.