ContactUsLinkComponent
Glove Guard® Belt Loop Glove Clip with Safety Breakaway, Magid
Glove Guard® Belt Loop Glove Clip with Safety Breakaway, Magid
Catalog # 32889-685
Supplier:  Magid Glove
Glove Guard® Belt Loop Glove Clip with Safety Breakaway, Magid
Catalog # 32889-685
Supplier:  Magid Glove
Supplier Number:  GG1939Y
Specifications
  • Glove color:
    Yellow
  • Description:
    Glove Guard Clip
  • Cat. No.:
    32889-685
  • Supplier no.:
    GG1939Y
Specifications
About this item
The glove clip with safety breakaway keep gloves readily accessible with glove guards glove clip glove fastener.
  • Clips onto belt loop, heavy waist band, or certain jacket pockets
  • Dielectric
  • Safety break-away capability
The glove guards patented safety breakaway helps prevent individuals from being pulled into machinery, and from being pulled off balance if gloves snag while climbing a ladder.