Order Entry
United States
ContactUsLinkComponent
Magid® Clear Vinyl Protective Sleeve, Magid Glove
Magid® Clear Vinyl Protective Sleeve, Magid Glove
Catalog # 46620-060
Supplier:  Magid Glove
Small Business Enterprise
Magid® Clear Vinyl Protective Sleeve, Magid Glove
Catalog # 46620-060
Supplier:  Magid Glove
Supplier Number:  19018
Small Business Enterprise

Specifications

  • Garment application:
    Industrial,
    Laboratory
  • Gender:
    Unisex
  • Length:
    45.7 cm (18")
  • Fabric:
    Vinyl
  • Thickness:
    8 mil
  • Cat. No.:
    46620-060
  • Supplier no.:
    19018

Specifications

About this item

This clear vinyl sleeve is completely heat-sealed and great for food processing, meat packing, sanitation and maintenance.

  • Heat sealed with no exposed elastic seams