Order Entry
United States
Orders LinkContactUsLinkComponent
 

Dihydroethidium

25 mg
Catalog # 103011-326
Marketsource
Supplier:   Anaspec
 :  AS-85704
Dihydroethidium
Catalog # 103011-326
Supplier:  Anaspec
Marketsource
CAS Number:  104821-25-2
Dihydroethidium

25 mg

CAS Number:  104821-25-2
Price
$431.15
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Bind to DNA/RNA (red fluorescence) upon oxidation. Store at -20C desiccated and protected from light

  • Spectral properties: Abs/Em = 518/605 nm (after oxidation)
  • Solvent system: DMSO
  • form: Dark red, pink Solid
Synonyms:  3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine, DHE, 5-Ethyl-6-phenyl-5,6-dihydrophenanthridine-3,8-diamine, 2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine
  • CAS Number:104821-25-2
  • Formula:C₂₁H₂₁N₃
  • MDL Number:MFCD00077335
  • Molecular Weight:315.412 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Size:
    25 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.