Magid® CutMaster® Gloves with Reinforced Thumb Crotch, Magid
Supplier: Magid Glove
Certificates
About this item
A 100% Kevlar, seamless, machine knit glove provides excellent all-purpose hand protection.
- Extra heavy-weight
- Reversible for longer wear
- Sewn-on, reinforced thumb crotch
Kevlar fiber is five times stronger than steel. This style includes a sewn-on thumb crotch for increased strength and durability in that most worn of areas.
The color-coded overedge wrist prevents unraveling and keeps a tight fit after washing. The Kevlar glove is a strong, cut-resistant extra heavy-weight glove that excels in any industry.
Certifications: ANSI Cut Level 4 protection.
Specifications
- Catalog No:
- 69100-539
- 69100-541
- 69100-545
- 69100-543
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable/Reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Material:
- Kevlar®
- Kevlar®
- Kevlar®
- Kevlar®