ContactUsLinkComponent
Magid® Nylon Mesh Sleeve, Magid Glove
  32893-183
Magid® Nylon Mesh Sleeve, Magid Glove
  32893-183
 :  230-7X13
  • Garment fit:
    Regular
  • Type:
    Magid® Nylon mesh black sleeve
  • Closure type:
    With Velcro
  • Garment application:
    Industrial,
    Laboratory
  • Gender:
    Unisex
  • Risk area:
    Cuts
  • Color:
    White
  • L×W:
    7×13"
  • Fabric:
    Nylon
  • Cat. No.:
    32893-183
  • Supplier no.:
    230-7X13
The Magid® Nylon Mesh Sleeve features a double layer nylon cane mesh construction for excellent cut and abrasion resistance.
  • Can be washed multiple times
The cut resistant sleeve is white nylon with Black Velcro closure for easy removal and size adjustment.
 : Meets ANSI Cut Level 4.