ContactUsLinkComponent
Magid® SparkGuard® Flame Resistant Bib Style Apron, Magid Glove
Magid® SparkGuard® Flame Resistant Bib Style Apron, Magid Glove
Catalog # 69100-053
Supplier:  Magid Glove
Magid® SparkGuard® Flame Resistant Bib Style Apron, Magid Glove
Catalog # 69100-053
Supplier:  Magid Glove
Supplier Number:  18040
Specifications
  • Garment application:
    Industrial
  • Risk area:
    Flame-retardant
  • Color:
    Green
  • L×W:
    30×25"
  • Fabric:
    Cotton
  • Cat. No.:
    69100-053
  • Supplier no.:
    18040
Specifications
About this item
Magid SparkGuard Flame Resistant Bib Style Apron provides flame resistance even after repeated laundering.
  • Overedge bib and bottom
  • Loose string ties
This protective apron features an overedge bib and bottom.
Certifications: Meets ASTM D6413-99 for flame resistance of textiles.