Platinum 0.8% on carbon powder wetted with water 23 µm for use as a catalyst, reduced , BASF Catalyst
Supplier: Spectrum Chemical Mfg. Corp.
Certificates
About this item
CAS Number:7440-06-4
:
Formula:Pt
MDL Number:MFCD00011179
:195.08 g/mol
Storage Temperature:Ambient
Frequently Bought Together