Statfree® Document Holder Protektive Pak
Static dissipative document holder, Vinyl, 10×12"
Catalog # 76277-264
Supplier: Menda
Supplier #: 47121
Jump to:
Product Details & Documents
Low charging and dissipative Statfree® Document Holders are used in static sensitive testing and production environments to hold work orders and drawings.
- Low charging and dissipative, provides path-to-ground
- Single opening
Imprinted with ESD protective symbol.
Packaging: Sold in packs of 25 each.
Specifications
- Description:Static dissipative document holder
- Dimensions:10×12"
- Material:Vinyl