ContactUsLinkComponent
Tech-Med® Graduated Beakers, DUKAL Corporation®
Tech-Med® Graduated Beakers, DUKAL Corporation®
Catalog # 76235-146
Supplier:  Dukal
Supplier #: 4244
undefined
Tech-Med® Graduated Beakers, DUKAL Corporation®
Catalog # 76235-146
Supplier:  Dukal
Supplier Number:  4244
Supplier #: 4244
Log in for availability
Select an Option
16 oz.
Select an Option
Stainless Steel Graduated Beaker
Select an Option
76235-146
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
About this item
Beakers are crafted without seams to prevent bacteria growth and allow for easy cleaning.
  • Made of the highest grade stainless steel (18/8)
  • Autoclavable
Frequently Bought Together