Multiple Affinity Removal Spin Cartridge - Hu-6HC (Human), Agilent Technologies
76204-890
: Avantor
- Capacity:0.45 ml
- Description:Multiple Affinity Removal Spin Cartridge - Hu-6HC (Human)
- Cat. No.:76204-890
The multiple affinity removal spin cartridge – Human 6 (standard or high-capacity) is designed to chromatographically remove six interfering high-abundant proteins from human plasma samples. Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
- Cartridge eliminates approximately 85 to 90% of total protein (albumin, IgG, IgA, transferrin, haptoglobin, and antitrypsin)
- Serum load volume ranges between 7 to 10 µl per run for the standard cartridge, up to 14 to 16 µl for the high-capacity cartridge
- Total load volume is 200 µl (serum plus buffer)
- Minimize sample loss by utilizing our rapid, single injection protocols and optimized buffers
- Process the most plasma/serum over the column lifetime, up to 2000 µl for the standard cartridge, 3200 µl for the high-capacity cartridge
Protocols and column are optimized for use with human serum. Column not recommended for use with mouse serum.Compatible with standard benchtop microcentrifuges and luer-lok syringes.