Peptide Cleanup C18 Spin Tubes, Agilent Technologies
76204-862
: Avantor
- Capacity:10 -150 µl
- Description:Peptide Cleanup Spin Tubes
- Cat. No.:76204-862
- Phase:C18
Peptide cleanup spin tubes are packed with C18 reverse phase resin used for purification and concentration of peptide, protein digests, proteins, or oligonucleotide samples by centrifuging sample through spin tube packing. Typical applications include sample preparation for analysis using ESI and MALDI-MS.
- Each tubes can process a digest from 20 ng of protein of 30 µg of total peptide
- Packed with 8 mg proprietery C18 resin
- Uses an easy-to-follow, 6-step protocol that enables purification of peptide samples in less than 30 minutes
Recommended one-time use only. Designed to be with standard benchtop microcentrifuge at 1500 g.
: Research use only.