ContactUsLinkComponent
Accessories for Stuart Digital Melting Point Apparatus, Cole Palmer
Accessories for Stuart Digital Melting Point Apparatus, Cole Palmer
Catalog # 76268-916
Supplier:  Antyila Scientific
undefined
Accessories for Stuart Digital Melting Point Apparatus, Cole Palmer
Catalog # 76268-916
Supplier:  Antyila Scientific
Supplier Number:  03013-64
Specifications
  • Description:
    SMP10/1 Glass Capillary Tubes, Open One End
  • Cat. No.:
    76268-916
  • Details:
    Soda glass Capillary Material; 3.94" (100 mm) Capillary Tube Length; 0.075" (1.9 mm) Capillary Tube Outer Diameter; 0.05" (1.3 mm) Capillary Tube Inner Diameter; 0.012" (0.3 mm) Capillary Tube Wall Thickness