Order Entry
United States
ContactUsLinkComponent
DNA Extraction, prepGEM Universal, MicroGEM
DNA Extraction, prepGEM Universal, MicroGEM
Supplier:  Avantor
Certificates
DNA Extraction, prepGEM Universal, MicroGEM
Supplier:  Avantor

Specifications

  • Method / Format:
    Extraction for direct PCR
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    prepGEM Universal

Specifications

Frequently Bought Together