Order Entry
United States
ContactUsLinkComponent
 
deconex® 11 UNIVERSAL, Cleaning Concentrate for Laboratory Glassware, TechnoTrade International
  MSPP-50010000K
deconex® 11 UNIVERSAL, Cleaning Concentrate for Laboratory Glassware, TechnoTrade International
  MSPP-50010000K
 :  500100.00-K05W

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    deconex® 11 UNIVERSAL
  • Size:
    5 l
  • Cat. No.:
    MSPP-50010000K
  • pH:
    11.8 (1% solution)
  • Form:
    Concentrate
  • Supplier no.:
    500100.00-K05W

 

 

deconex® 11 UNIVERSAL is a mildly alkaline, liquid concentrate with activated chlorine used for the cleaning and decontamination of laboratory glassware.

deconex® 11 UNIVERSAL exhibits a wide cleaning spectrum due to its composition. This allows even stubborn contaminants to be removed such as baked-on fats, oils, waxes, dried blood, proteins, residues of culture media, pen markings, pigments, traces of organic and inorganic materials and silicones.

Suitable for: Borosilicate glass 3.3, Duran®, quartz glass, porcelain, stainless steel, chrome nickel steel, polyethylene, polypropylene, Teflon®, silicone rubber, natural rubber.

Not suitable for: Aluminium, zinc, tin

For materials not mentioned please make your own specific compatibility tests or consult Borer Chemie AG