ELISA HRP Substrate, Lumi-Phos HRP, Lumigen
MSPP-PSA100
:
- Conjugate ELISA:HRP
- Description:Lumi-Phos HRP
- Size:100 ml
- Cat. No.:MSPP-PSA100
- Supplier no.:PSA-100
Lumi-Phos HRP (PS-atto) is a proprietary acridan based chemiluminescent substrate for ELISA detection of horseradish peroxidase (HRP) conjugated molecules. Lumi-Phos HRP couples a broad dynamic range with rapid generation of sustained luminescence.
- Excellent sensitivity - low picogram to femtogram
- Rapid peak intensity - reduced assay time and increased throughput
- Sustained luminescence - timing is less critical
- Temperature insensitive - analytical results are insensitive to temperatures from 22 to 35 °C reducing the need for precise temperature control
Reaction of the substrate with HRP enzyme rapidly generates sustained high-intensity chemiluminescence within seconds. Light emission is maximal at 450 nm.