Anti-Adgre1 Rat Monoclonal Antibody [clone: Recombinant CI:A3-1]
Catalog # 76285-134
Supplier: Enzo Life Sciences
Specifications
- Antibody Type:Primary
- Antigen Name:F4/80
- Antigen Symbol:Adgre1
- Clonality:Monoclonal
- Clone:Recombinant CI:A3-1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Size:200 µg
- Form:Liquid
- Antigen Synonyms:EMR1 receptor|EGF-like module receptor 1|Em Gpf4|Ly-71|Cell surface glycoprotein F4/80|TM7|EGF-like module
- UniProtKB:UniProt# Q61549
- Storage Buffer:In PBS containing 0.02% Proclin 300.
- Storage Temperature:–20 °C
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Thioglycollate stimulated peritoneal macrophages of mouse origin.
- Purification:Protein A affinity purified.
- Cat. No.:76285-134
Specifications
About this item
Recombinant monclonal antibody (Rat IgG2bκ).
Recommended Applications: Flow Cytometry, IHC
Type: Primary
Antigen: Adgre1
Clonality: Monoclonal
Clone: Recombinant CI:A3-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2bκ
Reactivity: Mouse