Anti-His Tag Mouse Monoclonal Antibody [clone: 3D5]
Catalog # 76285-058
Supplier: Enzo Life Sciences
Specifications
- Antibody Type:Primary
- Antigen Symbol:His Tag
- Clonality:Monoclonal
- Clone:3D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Western Blot:Yes
- Size:200 µg
- Form:Liquid
- Antigen Synonyms:Polyhistidine tag
- Storage Buffer:In PBS containing 0.02% Proclin 300.
- Storage Temperature:–20 °C
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Purified his tagged Fab fragment.
- Purification:Protein A affinity purified.
- Cat. No.:76285-058
Specifications
About this item
Recombinant monclonal antibody (Mouse IgG2aκ).
Recommended Applications: ELISA, WB
Type: Primary
Antigen: His Tag
Clonality: Monoclonal
Clone: 3D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2aκ
Reactivity: